Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion
Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion
Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion
Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion
Thermo Scientific Chemicals

Titanium(IV) oxide, 20-36% in H{2}O colloidal dispersion

UV protection in clear coatings | CAS: 13463-67-7 | O2Ti | 79.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
Catalog number 044517.36
also known as 044517-36
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS13463-67-7
IUPAC Namedioxotitanium
Molecular FormulaO2Ti
InChI KeyGWEVSGVZZGPLCZ-UHFFFAOYSA-N
SMILESO=[Ti]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormDispersion
Assay (special)Solids: 20-36%
UV protection in clear coatings. Photo electrodes of varying properties can be made using colloidal suspensions of titanium(IV) oxide nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only