Calcium D-gluconate gel, 2.5% w/w aq. soln.
Calcium D-gluconate gel, 2.5% w/w aq. soln.
Calcium D-gluconate gel, 2.5% w/w aq. soln.
Thermo Scientific Chemicals

Calcium D-gluconate gel, 2.5% w/w aq. soln.

CAS: 299-28-5 | C12H22CaO14 | 430.372 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
30 g
60 g
Catalog number 044542.15
Price (USD)
126.65
Special offer
Online exclusive
Ends: 28-Dec-2026
149.00
Save 22.35
Each
Quantity:
30 g
Request bulk or custom format
Price (USD)
126.65
Special offer
Online exclusive
Ends: 28-Dec-2026
149.00
Save 22.35
Each
Chemical Identifiers
CAS299-28-5
IUPAC Namecalcium bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate)
Molecular FormulaC12H22CaO14
InChI KeyNEEHYRZPVYRGPP-IYEMJOQQSA-L
SMILES[Ca++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)white to off-white
FormGel
Assay (Titration)>/= 2.3%
Calcium D-gluconate gel is used to treat hydrofluoric acid burns. It is used as a cardioprotective agent in hyperkalemia, antidote for black widow spider envenomation and an antidote for magnesium sulfate toxicity. It is also involved in the treatment of hypocalcemia. Further, it is used as skin conditioning as well as provides the source of calcium ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only