Europium rod, 9.52 mm (0.375in) dia, 99.9% (metals basis)
Europium rod, 9.52 mm (0.375in) dia, 99.9% (metals basis)
Europium rod, 9.52 mm (0.375in) dia, 99.9% (metals basis)
Thermo Scientific Chemicals

Europium rod, 9.52 mm (0.375in) dia, 99.9% (metals basis)

CAS: 7440-53-1 | Eu | 151.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mm
50 mm
Catalog number 044545.DJ
also known as 044545-DJ
Price (USD)
1,676.65
Online Exclusive
1,863.00
Save 186.35 (10%)
Each
Quantity:
50 mm
Request bulk or custom format
Price (USD)
1,676.65
Online Exclusive
1,863.00
Save 186.35 (10%)
Each
Chemical Identifiers
CAS7440-53-1
IUPAC Nameeuropium
Molecular FormulaEu
InChI KeyOGPBJKLSAFTDLK-UHFFFAOYSA-N
SMILES[Eu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey/silver
FormRod, 9.52 mm diameter
Total Metal Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged in mineral oil
RUO – Research Use Only