Copper wire, 1.5mm (0.06in) dia, Puratronic™, 99.999% (metals basis)
Copper wire, 1.5mm (0.06in) dia, Puratronic™, 99.999% (metals basis)
Copper wire, 1.5mm (0.06in) dia, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Copper wire, 1.5mm (0.06in) dia, Puratronic™, 99.999% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
5 m
Catalog number 044550.G1
also known as 044550-G1
Price (USD)
129.65
Special offer
Online exclusive
Ends: 28-Dec-2026
152.00
Save 22.35
Each
Quantity:
1 m
Request bulk or custom format
Price (USD)
129.65
Special offer
Online exclusive
Ends: 28-Dec-2026
152.00
Save 22.35
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 1.5mm (0.06in) diameter.
Appearance (Color)Brown
Assay from Supplier's CofA≥99.999% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only