4-Methyldiphenylamine, 98%
4-Methyldiphenylamine, 98%
4-Methyldiphenylamine, 98%
Thermo Scientific Chemicals

4-Methyldiphenylamine, 98%

CAS: 620-84-8 | C13H13N | 183.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 044560.09
also known as 044560-09
Price (USD)
61.65
Online exclusive
68.00
Save 6.35
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
61.65
Online exclusive
68.00
Save 6.35
Each
Specifications
CAS620-84-8
Chemical Name or Material4-Methyldiphenylamine
ColorBrown
EINECS Number210-655-6
Flash Point155°C (311°F)
View more
4-Methyldiphenylamine is used as a syntheses material intermediates as well as OLED materials. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only