Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol
Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol
Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol
Thermo Scientific Chemicals

Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol

CAS: 38218-24-5 | C9H21InO3 | 292.082 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number 044562.AE
also known as 044562-AE
Price (USD)
486.65
Special offer
Online exclusive
Ends: 30-Jun-2026
572.00
Save 85.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
486.65
Special offer
Online exclusive
Ends: 30-Jun-2026
572.00
Save 85.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS38218-24-5
Chemical Name or MaterialIndium (III) isopropoxide
ColorColorless
Concentration or Composition (by Analyte or Components)5% w/v isopropanol
View more
Precursor used in the preparation of indium oxide containing films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only