Aluminum etchant
Thermo Scientific Chemicals

Aluminum etchant

Standard Aluminum Etchant For Use On Silicon Devices and Other Microelectronic Applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 qt.
1 gal.
Catalog number 044581.LV
also known as 044581-LV
Price (USD)
283.00
Each
Quantity:
1 gal.
Request bulk or custom format
Price (USD)
283.00
Each
Specifications
Chemical Name or MaterialAluminum etchant
DOT InformationDOT Class: 8, Packaging Group: II
Health Hazard 1H290-H314-H335
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
MDL NumberMFCD08064399
View more
Standard aluminum etchant for use on silicon devices and other microelectronic applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

What is the solubility of Aluminum etchant?

Aluminum etchant is miscible with water.

What is the flash point of Aluminum etchant?

Aluminum etchant is non-inflammable.

What is the specific gravity (25 degrees C) of Aluminum etchant?

The specific gravity (25 degrees C) of Aluminum etchant is 1.45.

What is the freezing point of Aluminum etchant?

The freezing point of Aluminum etchant is <0 degrees C.

What is the boiling point of Aluminum etchant?

The boiling point of Aluminum etchant is >100 degrees C.