What is the etch rate of Copper etchant at 40 degrees C?
Copper etchant has an etch rate of 1 mil/minute at 40 degrees C (etches 1 oz copper in 90 seconds).
Search
View additional product information for Copper etchant - FAQs (044583.LV, 044583.R0)
5 product FAQs found