Gold etchant - FAQs
Gold etchant - FAQs
View additional product information for Gold etchant - FAQs (044584.LV, 044584.R0)
5 product FAQs found
What is the etch rate of Gold etchant at 25 degrees C?
Gold etchant has an etch rate of 28 Å/sec at 25 degrees C.
How do I reduce undercutting of an etchant?
Undercutting of the etchant can be reduced by increasing the rate of stirring or agitation.
Do I need to dilute an etchant?
No, it is a ready-to-use solution. Dilution will only be necessary if you want to reduce the etch rate.
How do I reduce the etch rate of an etchant?
The etch rate of the etchant can be reduced by adding 1 part deionized water to 2 parts etchant. This will reduce the etch rate approximately 50%.
How do I increase the etch rate of an etchant?
The etch rate of the etchant will be increased by increasing the rate of stirring or agitation.