Neodymium(III) acetate hydrate, 99.9% (REO)
Neodymium(III) acetate hydrate, 99.9% (REO)
Neodymium(III) acetate hydrate, 99.9% (REO)
Neodymium(III) acetate hydrate, 99.9% (REO)
Thermo Scientific Chemicals

Neodymium(III) acetate hydrate, 99.9% (REO)

CAS: 334869-71-5 | C6H9NdO6 | 321.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 044587.30
also known as 044587-30
Price (USD)
253.65
Special offer
Online exclusive
Ends: 30-Jun-2026
298.00
Save 44.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
253.65
Special offer
Online exclusive
Ends: 30-Jun-2026
298.00
Save 44.35 (15%)
Each
Chemical Identifiers
CAS334869-71-5
IUPAC Nameneodymium(3+) triacetate
Molecular FormulaC6H9NdO6
InChI KeyKETUDCKOKOGBJB-UHFFFAOYSA-K
SMILES[Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Neodymium Acetate, mainly used for glass, crystal and capacitors. It is useful in protective lenses for welding goggles. It is also used in CRT displays to enhance contrast between reds and greens. It is highly valued in glass manufacturing for its attractive purple coloring to glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only