Ammonium tungsten oxide, 99.99+% (metals basis)
Ammonium tungsten oxide, 99.99+% (metals basis)
Ammonium tungsten oxide, 99.99+% (metals basis)
Ammonium tungsten oxide, 99.99+% (metals basis)
Thermo Scientific Chemicals
Ammonium tungsten oxide, 99.99+% (metals basis)
CAS: 11140-77-5 | H40N10O41W12 | 3042.429 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 044588.18
also known as 044588-18
Price (USD)
387.65
Online Exclusive
430.00
Save 42.35 (10%)
Each
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)
387.65
Online Exclusive
430.00
Save 42.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS11140-77-5
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
Ammonium Paratungstate is used as a source for high-purity tungsten oxides, tungsten metal powders, carbides, or as a laboratory reagent. Other applications are in the fields of absorbent gels, coloring agent in the porcelain industry, and the catalyst industry.