HSG, Heat sink grease
Thermo Scientific Chemicals

HSG, Heat sink grease

thermally Conductive Heat Sink Grease Applied To the Base and Mounting Stud of Transistors, Diodes and Rectifiers, As a Coupling Agent
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 oz.
16 oz.
Catalog number 044589.3D
also known as 044589-3D
Price (USD)
194.00
Each
Quantity:
16 oz.
Request bulk or custom format
Price (USD)
194.00
Each
Specifications
Chemical Name or MaterialHSG
ColorWhite
Density2.3
Health Hazard 1H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 2In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
View more
Thermally conductive heat sink grease applied to the base and mounting stud of transistors, diodes and rectifiers, as a coupling agent, to reduce thermal contact resistance. Ideal for sputtering target applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only