Titanium(IV) 2-ethylhexanoate, 97%
Titanium(IV) 2-ethylhexanoate, 97%
Titanium(IV) 2-ethylhexanoate, 97%
Thermo Scientific Chemicals

Titanium(IV) 2-ethylhexanoate, 97%

CAS: 3645-34-9 | C32H60O8Ti | 620.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 044602.22
also known as 044602-22
Price (USD)
254.65
Special offer
Online exclusive
Ends: 30-Jun-2026
299.00
Save 44.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
254.65
Special offer
Online exclusive
Ends: 30-Jun-2026
299.00
Save 44.35 (15%)
Each
Chemical Identifiers
CAS3645-34-9
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (Titration)≥96.0%
Appearance (Color)Dark yellow to orange
Titanium(IV) 2-ethylhexanoate acts as a coupling agent to improve the adhesion of the fiber and resin in Kevla-phenolic composites. It is used as a catalyst in the polymerization of cyclic poly(butylene terephthalate) oligomers for composite applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only