(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine
Thermo Scientific Chemicals

(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine

Asymmetric hydrogenation | CAS: 442905-33-1 | C46H50N2O4P2 | 756.864 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.1 g
0.5 g
Catalog number 044613.77
also known as 044613-77
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Quantity:
0.1 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.10
Save 12.45 (15%)
Each
Specifications
CAS442905-33-1
Chemical Name or Material(R)- (+)-2,2',6,6'-Tetramethoxy-4,4'-bis (di (3,5-xylyl)phosphino)-3,3'-bipyridine
Formula Weight756.86
IUPAC Name[3-[4-bis(3,5-dimethylphenyl)phosphanyl-2,6-dimethoxypyridin-3-yl]-2,6-dimethoxypyridin-4-yl]-bis(3,5-dimethylphenyl)phosphane
InChI KeyJRTHAKOHBMETRC-UHFFFAOYSA-N
View more
(R)-(+)-2,2',6,6'-Tetramethoxy-4,4'-bis(di(3,5-xylyl)phosphino)-3,3'-bipyridine is used in asymmetric hydrogenation.It acts as a ligand used in the asymmetic hydrogenation of β-keto esters, 2-arylacrylates, aryl ketones , and other substrates. Other applications include it is used widely as a Catalyst, when combined with Rh, Ru and Ir precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only