Buffer HF-Improved, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer HF-Improved, Thermo Scientific Chemicals

Used For Pretreatment of Planar Silicon Devices When Plating With Nickel Plating Solution Products 44069 and 44070
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 044627.K2
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Specifications
Boiling Point103°C to 105°C
Chemical Name or MaterialBuffer HF-Improved
ColorColorless
DOT InformationDOT Class: 8; Packaging Group: II
Density1.12 g/cm3 at 20°C
View more
Used for pretreatment of planar silicon devices when plating with Nickel plating solution products 44069 and 44070.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only