Silicon(IV) 2-ethylhexanoate, nominally 75% in ethanol
Silicon(IV) 2-ethylhexanoate, nominally 75% in ethanol
Silicon(IV) 2-ethylhexanoate, nominally 75% in ethanol
Thermo Scientific Chemicals

Silicon(IV) 2-ethylhexanoate, nominally 75% in ethanol

CAS: 67939-81-5 | C32H60O8Si | 600.909 g/mol
Have Questions?
Catalog NumberQuantity
044635.30
also known as 044635-30
250 g
Catalog number 044635.30
also known as 044635-30
Price (USD)
487.65
Special offer
Online exclusive
Ends: 30-Jun-2026
573.00
Save 85.35 (15%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
487.65
Special offer
Online exclusive
Ends: 30-Jun-2026
573.00
Save 85.35 (15%)
Each
Specifications
CAS67939-81-5
Chemical Name or MaterialSilicon (IV) 2-ethylhexanoate
ColorYellow
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHANOL SOLUTIONS
Formula Weight600.91
View more
In the manufacture of its esters for artificial flavors, and in the manufacture of hexyl derivatives, such as hexylphenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For Manufacturing and Laboratory Use. Not for beverage use