Cerium(IV) oxide, 99.5% (REO)
Cerium(IV) oxide, 99.5% (REO)
Cerium(IV) oxide, 99.5% (REO)
Thermo Scientific Chemicals

Cerium(IV) oxide, 99.5% (REO)

CAS: 1306-38-3 | CeO2 | 172.114 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 044662.14
also known as 044662-14
Price (USD)
69.65
Online Exclusive
77.60
Save 7.95 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
77.60
Save 7.95 (10%)
Each
Chemical Identifiers
CAS1306-38-3
IUPAC Namedioxocerium
Molecular FormulaCeO2
InChI KeyCETPSERCERDGAM-UHFFFAOYSA-N
SMILESO=[Ce]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.5% max.
Particle Size50nm
Cerium(IV) oxide is used in ceramics, and to polish glass and stones. It is applied as solid electrolyte in fuel cells. It can also be employed as a co-catalyst in many number of reactions. It is also used as an antioxidant, and to generate hydrogen through water splitting. It is used as a high-temperature hydrocarbon oxidation catalyst in self-cleaning ovens. It can be used in infrared filters, incandescent mantles, and sunscreens in combination with other materials. It finds use as a catalyst support, and to sensitize photosensitive glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only