Soda lime, indicating, ACS - FAQs
Soda lime, indicating, ACS - FAQs
View additional product information for Soda lime, indicating, ACS - FAQs (044697.36, 044697.A4)
2 product FAQs found
Does Soda lime, indicating, ACS desiccant have an approved regeneration process?
Soda lime, indicating, ACS cannot be regenerated.
What colors indicate fresh, and fully spent states of Soda lime, indicating, ACS?
When Soda lime, indicating, ACS is fresh, it should appear white to light grey. As the product is consumed, it will eventually turn violet when fully spent.