Phenyl phosphate disodium salt hydrate, 98%
Phenyl phosphate disodium salt hydrate, 98%
Phenyl phosphate disodium salt hydrate, 98%
Phenyl phosphate disodium salt hydrate, 98%
Thermo Scientific Chemicals

Phenyl phosphate disodium salt hydrate, 98%

Used in phosphatase determinations | CAS: 3279-54-7 | C6H5Na2O4P | 218.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 044711.22
also known as 044711-22
Price (USD)
306.00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
306.00
Each
Add to cart
Chemical Identifiers
CAS3279-54-7
IUPAC Namedisodium phenyl phosphate
Molecular FormulaC6H5Na2O4P
InChI KeyTYJOJLOWRIQYQM-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]P([O-])(=O)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5%
FormCrystalline powder
Water Content (Karl Fischer Titration)11 - 17%
Assay (HPLC)≥97.5%
Phenyl phosphate disodium salt is used in phosphatase determinations. It is also used as reagent in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only