Magnesium oxide, nanopowder, 99+% (metals basis)
Magnesium oxide, nanopowder, 99+% (metals basis)
Magnesium oxide, nanopowder, 99+% (metals basis)
Thermo Scientific Chemicals

Magnesium oxide, nanopowder, 99+% (metals basis)

S.A. >7.3m2/g | CAS: 1309-48-4 | MgO | 40.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 044733.14
also known as 044733-14
Price (USD)
70.10
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
70.10
Each
Chemical Identifiers
CAS1309-48-4
IUPAC Nameoxomagnesium
Molecular FormulaMgO
InChI KeyCPLXHLVBOLITMK-UHFFFAOYSA-N
SMILESO=[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Total Metal Impurities≤1%
Surface Area≥30m²/g
Appearance (Color)White
Particle Size40-60nm
Magnesium oxide is used as a laboratory reagent and an insulator in industrial cables. It acts as a reference material for calorimetry. It is widely used as a basic refractory material for crucibles, as a protective coating in plasma displays, as an oxide barrier in spin-tunneling devices, as an optical material, as a component of tubular construction in heating elements and also in libraries for preserving books. Magnesium oxide microspheres are used in the enrichment method of polycyclic aromatic hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only