Search
View additional product information for Silicon oxide, catalyst support, high surface area - FAQs (044740.22, 044740.36, 044740.A3)
1 product FAQs found
Silicon oxide, catalyst support, high surface area comes in pellet form.