Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000
Thermo Scientific Chemicals

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 63,000

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
4 L
Catalog number 044764.AK
also known as 044764-AK
Price (USD)
63.90
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (USD)
63.90
Each
Add to cart
Chemical Identifiers
CAS9016-00-6
Molecular Formula(C2H6OSi)n
SMILESC[Si](C)(-*)O-*
MDL NumberMFCD00084411
SpecificationsSpecification SheetSpecification Sheet
Formliquid
Viscosity9000 to 11000 cSt at 25 deg c
Appearance (Color)Clear, colorless
Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppresses the formation of foams and is a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only