Molybdenum etchant, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum etchant, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
044768.LV
also known as 044768-LV
1 gal.
Catalog number 044768.LV
also known as 044768-LV
Price (USD)
486.65
Online Exclusive
541.00
Save 54.35 (10%)
Each
Quantity:
1 gal.
Request bulk or custom format
Price (USD)
486.65
Online Exclusive
541.00
Save 54.35 (10%)
Each
Specifications
Chemical Name or MaterialMolybdenum etchant
ColorAmber
DOT InformationDOT Class: 8, Packaging Group: III
Density1.2
Health Hazard 1H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the etch rate of Molybdenum etchant at 30 degrees C and 80 degrees C?

Molybdenum etchant has an etch rate of 55 Å/sec at 30 degrees C and an etch rate of 85 Å/sec at 80 degrees C.

How do I reduce undercutting of an etchant?

Undercutting of the etchant can be reduced by increasing the rate of stirring or agitation.

Do I need to dilute an etchant?

No, it is a ready-to-use solution. Dilution will only be necessary if you want to reduce the etch rate.

How do I reduce the etch rate of an etchant?

The etch rate of the etchant can be reduced by adding 1 part deionized water to 2 parts etchant. This will reduce the etch rate approximately 50%.

How do I increase the etch rate of an etchant?

The etch rate of the etchant will be increased by increasing the rate of stirring or agitation.