Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zirconium(IV) n-propoxide, 70% w/w in n-propanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044770.18
also known as 044770-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS23519-77-9
IUPAC Nametetrapropoxyzirconium
Molecular FormulaC12H28O4Zr
InChI KeyXPGAWFIWCWKDDL-UHFFFAOYSA-N
SMILESCCCO[Zr](OCCC)(OCCC)OCCC
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Suppliers CofAZrO2 Content: 27.0-29.0%
Specific Gravity1.050-1.090 g/mL
Zirconium(IV) n-propoxide is used as a precursor for the preparation of Zirconia. It is used as coupling agent and petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
RUO – Research Use Only