Allyl cyanide, 97%
Allyl cyanide, 97%
Allyl cyanide, 97%
Thermo Scientific Chemicals

Allyl cyanide, 97%

CAS: 109-75-1 | C4H5N | 67.091 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 044775.22
also known as 044775-22
Price (USD)
164.65
Special offer
Online exclusive
Ends: 30-Jun-2026
193.00
Save 28.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
164.65
Special offer
Online exclusive
Ends: 30-Jun-2026
193.00
Save 28.35 (15%)
Each
Chemical Identifiers
CAS109-75-1
IUPAC Namebut-3-enenitrile
Molecular FormulaC4H5N
InChI KeySJNALLRHIVGIBI-UHFFFAOYSA-N
SMILESC=CCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Assay from Supplier's CofA≥96.0%
Allyl cyanide is used as an anti-microbial. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only