Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)
Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)
Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)
Thermo Scientific Chemicals

Silicon(IV) oxide, amorphous, nanopowder, 99.9% (metals basis)

CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 044781.22
also known as 044781-22
Price (USD)
180.65
Online Exclusive
200.00
Save 19.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
180.65
Online Exclusive
200.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to gray
Total Metal Impurities≤0.1%
CommentParticle Size, approx.: 20 nm
FormPowder
Silicon(IV) oxide is used as sodium aluminate stabilizer. It is also used as SDS mixture of sodium alkyl sulfates consisting chiefly of sodium lauryl sulfate. It is also acts as chain transfer agent in polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only