Soda lime, indicating
Soda lime, indicating
Soda lime, indicating
Thermo Scientific Chemicals

Soda lime, indicating

CAS: 8006-28-8 | CaHNaO2 | 96.074 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Catalog number 044786.36
also known as 044786-36
Price (USD)
146.65
Special offer
Online exclusive
Ends: 28-Dec-2026
172.00
Save 25.35
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
146.65
Special offer
Online exclusive
Ends: 28-Dec-2026
172.00
Save 25.35
Each
Chemical Identifiers
CAS8006-28-8
IUPAC Namesodium oxocalcium hydroxide
Molecular FormulaCaHNaO2
InChI KeyHUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES[OH-].[Na+].O=[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
CommentCO₂ absorption capacity 19.0% min
Impurity contentFines 1% max
Loss on Dryingat 200? 7% max
Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only