(Perfluorocyclohexyl)methyl acrylate, 96%, stab. with 50ppm to 100ppm 4-methoxyphenol
(Perfluorocyclohexyl)methyl acrylate, 96%, stab. with 50ppm to 100ppm 4-methoxyphenol
(Perfluorocyclohexyl)methyl acrylate, 96%, stab. with 50ppm to 100ppm 4-methoxyphenol
Thermo Scientific Chemicals

(Perfluorocyclohexyl)methyl acrylate, 96%, stab. with 50ppm to 100ppm 4-methoxyphenol

CAS: 40677-94-9 | C10H5F11O2 | 366.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 044793.14
also known as 044793-14
Price (USD)
445.65
Online Exclusive
495.00
Save 49.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
445.65
Online Exclusive
495.00
Save 49.35 (10%)
Each
Chemical Identifiers
CAS40677-94-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Assay (GC)≥ 95.0%
FormLiquid
Commentstabilized with 50-100ppm 4-methoxyphenol
(Perfluorocyclohexyl)methyl acrylate is used in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only