Ethyl carbamate, 98%
Ethyl carbamate, 98%
Ethyl carbamate, 98%
Thermo Scientific Chemicals

Ethyl carbamate, 98%

CAS: 51-79-6 | C3H7NO2 | 89.094 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 044804.18
also known as 044804-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS51-79-6
IUPAC Nameethyl carbamate
Molecular FormulaC3H7NO2
InChI KeyJOYRKODLDBILNP-UHFFFAOYSA-N
SMILESCCOC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.5%
FormCrystalline powder
Assay from Supplier's CofAChloride: ≤0.01%
Assay (GC)≥97.5%
Melting Point46-52?C
View more

Ethyl carbamate in its molten state is good solvent for various organic materials, preparation & modification of amino resins, as solubilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only