Cyclopentane, HPLC Grade
Cyclopentane, HPLC Grade
Cyclopentane, HPLC Grade
Thermo Scientific Chemicals

Cyclopentane, HPLC Grade

For HPLC and Spectrophotometry | CAS: 287-92-3 | C5H10 | 70.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
4 L
Catalog number 044814.AK
also known as 044814-AK
Price (USD)
54.50
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
54.50
Each
Chemical Identifiers
CAS287-92-3
IUPAC Namecyclopentane
Molecular FormulaC5H10
InChI KeyRGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILESC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>/= 99.8% as sum of C5 + C6 hydrocarbons
Residual water0.02%
Optical Absorbance at210 nm 0.50 AU, max
Residue on Evaporation3ppm
FormClear liquid
View more
Cyclopentane is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only