Mercury(II) bromide, ACS
Mercury(II) bromide, ACS
Mercury(II) bromide, ACS
Thermo Scientific Chemicals

Mercury(II) bromide, ACS

CAS: 7789-47-1 | Br2Hg | 360.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 044816.14
also known as 044816-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS7789-47-1
IUPAC Namemercury(2+) dibromide
Molecular FormulaBr2Hg
InChI KeyNGYIMTKLQULBOO-UHFFFAOYSA-L
SMILES[Br-].[Br-].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCl 0.25%
Impurity contentIf a Mercury (I) salt is present the product darkens on exposure to light.
CommentResidue after reduction 0.02%
Appearance (Color)White or having at most a very faint yellow tinge
Impurity contentInsoluble in methanol 0.05%
Mercury(II) bromide is used as a reagent in the Koenigs-Knorr reaction, which forms glycoside linkages on carbohydrates. It is also used to test for the presence of arsenic, as recommended by the Pharmacopoeia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only