Dichlorobis(tricyclohexylphosphine)palladium(II)
Dichlorobis(tricyclohexylphosphine)palladium(II)
Dichlorobis(tricyclohexylphosphine)palladium(II)
Thermo Scientific Chemicals

Dichlorobis(tricyclohexylphosphine)palladium(II)

CAS: 29934-17-6 | C36H66Cl2P2Pd | 738.19 g/mol
Have Questions?
Catalog NumberQuantity
044844.03
also known as 044844-03
1 g
Catalog number 044844.03
also known as 044844-03
Price (USD)
161.65
Special offer
Online exclusive
Ends: 30-Jun-2026
190.00
Save 28.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
161.65
Special offer
Online exclusive
Ends: 30-Jun-2026
190.00
Save 28.35 (15%)
Each
Specifications
CAS29934-17-6
Chemical Name or MaterialDichlorobis (tricyclohexylphosphine)palladium (II)
Concentration or Composition (by Analyte or Components)Pd 14.4%
Formula Weight738.18
Health Hazard 1H302-H315-H319-H335
View more
It is used as catalyst for C-C and C-N coupling reaction. It is used in the homogeneous carbonylation of chloroarenes under mild conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only