Tricyclohexylphosphine, technical grade
Tricyclohexylphosphine, technical grade
Tricyclohexylphosphine, technical grade
Thermo Scientific Chemicals

Tricyclohexylphosphine, technical grade

CAS: 2622-14-2 | C18H33P | 280.436 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 044854.06
also known as 044854-06
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.00
Save 11.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.00
Save 11.35 (15%)
Each
Chemical Identifiers
CAS2622-14-2
IUPAC Nametricyclohexylphosphane
Molecular FormulaC18H33P
InChI KeyWLPUWLXVBWGYMZ-UHFFFAOYSA-N
SMILESC1CCC(CC1)P(C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Formsolid
Assay (GC)> 88.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only