Sponge Nickel, A-4000, promoted with 2% Iron and 2.5% Chromium, Thermo Scientific Chemicals
Sponge Nickel, A-4000, promoted with 2% Iron and 2.5% Chromium, Thermo Scientific Chemicals
Sponge Nickel, A-4000, promoted with 2% Iron and 2.5% Chromium, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sponge Nickel, A-4000, promoted with 2% Iron and 2.5% Chromium, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 044866.22
also known as 044866-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Specifications
AppearanceDark gray sponge
CAS7697-37-2
ChEBICHEBI:48107
Chemical Name or MaterialNickel
ColorGray
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only