Zinc oxide, NanoTek∣r
Zinc oxide, NanoTek∣r
Zinc oxide, NanoTek∣r
Zinc oxide, NanoTek∣r
Thermo Scientific Chemicals

Zinc oxide, NanoTek∣r

CAS: 1314-13-2 | OZn | 81.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 044899.36
also known as 044899-36
Price (USD)
137.65
Special offer
Online exclusive
Ends: 30-Jun-2026
162.00
Save 24.35 (15%)
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)
137.65
Special offer
Online exclusive
Ends: 30-Jun-2026
162.00
Save 24.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS1314-13-2
IUPAC Nameoxozinc
Molecular FormulaOZn
InChI KeyXLOMVQKBTHCTTD-UHFFFAOYSA-N
SMILESO=[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Particle Size45-89 nm (average particle size)
Surface Area12-24 m²/g (specific surface area)
FormPowder
NanoTek™ zinc oxide, due to the small particle size, can provide UV protection while maintaining a high level of transparency in coatings, polymers, caulks, adhesives and other resin systemsZinc oxide nanoparticles are used in the manufacture of rubber and cigarettes. It plays an important role in paints as a coating agent. It is used in creams and ointments, which are used to treat skin diseases. It acts as an additive in the manufacture of concrete. It is also used in ceramic industry and as an additive in food products such as breakfast cereals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only