ICP-MS General Memory & Interference Check Sample Mix for 6020, Specpure™
Thermo Scientific Chemicals

ICP-MS General Memory & Interference Check Sample Mix for 6020, Specpure™

Have Questions?
Catalog NumberQuantity
044935.AE
also known as 044935-AE
100 mL
Catalog number 044935.AE
also known as 044935-AE
Price (USD)
182.65
Special offer
Online exclusive
Ends: 30-Jun-2026
215.00
Save 32.35 (15%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
182.65
Special offer
Online exclusive
Ends: 30-Jun-2026
215.00
Save 32.35 (15%)
Each
Specifications
Chemical Name or MaterialICP-MS General Memory & Interference Check Sample Mix for 6020
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1H290-H315-H319
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only