Nitinol foil, 0.25mm (0.01in) thick, superelastic, flat annealed, pickled surface
Thermo Scientific Chemicals

Nitinol foil, 0.25mm (0.01in) thick, superelastic, flat annealed, pickled surface

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 044953.FF
also known as 044953-FF
Price (USD)
131.65
Online Exclusive
146.00
Save 14.35 (10%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
131.65
Online Exclusive
146.00
Save 14.35 (10%)
Each
Specifications
Chemical Name or MaterialNitinol foil
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL NumberMFCD02091734
View more
Nitinol foil is used in photo chemically-etched parts, formed sheet shapes, EDM or laser cut shapes, cell-phone technology as a retractable antenna, or microphone boom.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only