Nitinol foil, 0.38mm (0.015in) thick, superelastic, flat annealed,pickled surface
Thermo Scientific Chemicals

Nitinol foil, 0.38mm (0.015in) thick, superelastic, flat annealed,pickled surface

Form: Approx.1.56g/25 x 25mm; Af Approx. 10deg.C; Superelastic; Pickled surface
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 044954.FF
also known as 044954-FF
Price (USD)
72.65
Online exclusive
80.70
Save 8.05
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
72.65
Online exclusive
80.70
Save 8.05
Each
Specifications
AppearanceSilver-gray foil
Chemical Name or MaterialNitinol
ColorSilver to Gray
Health Hazard 1H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.
Health Hazard 2Wear suitable protective clothing and gloves. This material and/or its container must be disposed of as hazardous waste.
View more
Nitinol foil is used in photo chemically-etched parts, formed sheet shapes, EDM or laser cut shapes, cell-phone technology as a retractable antenna, or microphone boom.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only