Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)
Thermo Scientific Chemicals

Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II)

CAS: 16971-33-8 | C55H46ClOP3Ru | 952.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 044955.03
also known as 044955-03
Price (USD)
109.65
Online Exclusive
122.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
109.65
Online Exclusive
122.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS16971-33-8
IUPAC Namechloro(hydrido)rutheniumiumylium; methanidylidyneoxidanium; tris(triphenylphosphane)
Molecular FormulaC55H46ClOP3Ru
InChI KeyIMPHYUNMDWWJAH-UHFFFAOYSA-M
SMILESCl[RuH++].[C-]#[O+].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)3.20-4.20% Chlorine
Assay (unspecified)4.30-5.50% Hydrogen
Assay (unspecified)68.40-70.40% Carbon
Carbonylchlorohydridotris(triphenylphosphine)ruthenium(II) is used as a raw material to synthesize other ru-hydride catalysts. It is involved in the hydrogenation of ring-opening metathesis polymers such as norbornene and other cyclic olefin ROMP polymers used as plastics, resins and optical materials. It can act as an alternative catalyst to rhodium hydrogenation catalyst. It also serves as a catalyst in the hydrogenation of conjugated diene copolymers like acrylonitrile-butadiene rubber (NBR).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only