Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II), Pd 17.6%
Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II), Pd 17.6%
Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II), Pd 17.6%
Thermo Scientific Chemicals

Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II), Pd 17.6%

CAS: 29964-62-3 | C28H28Cl2P2Pd | 603.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 044971.03
also known as 044971-03
Price (USD)
197.65
Online Exclusive
219.00
Save 21.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
197.65
Online Exclusive
219.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS29964-62-3
IUPAC Name[4-(diphenylphosphanyl)butyl]diphenylphosphane; dichloropalladium
Molecular FormulaC28H28Cl2P2Pd
InChI KeyJQXJBXVWVPVTOO-UHFFFAOYSA-L
SMILESCl[Pd]Cl.C(CCP(C1=CC=CC=C1)C1=CC=CC=C1)CP(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)17.6% Pd min
Dichloro[bis(1,4-diphenylphosphino)butane]palladium(II) is used for the catalysis of styrene carbonylation, coupling of alkyl Grignard reagents with organic halides, selective monoalkylation of organic polyhalides, and modification of the dihalovinyl moiety of synthetic pyrethroids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only