Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 316
Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 316
Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 316
Thermo Scientific Chemicals

Stainless Steel gauze, 200 mesh woven from 0.05mm (0.002in) dia wire, Type 316

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 x 10 cm
20 x 30 cm
30 x 60 cm
Catalog number 045002.RU
also known as 045002-RU
Price (USD)
287.65
Online Exclusive
319.00
Save 31.35 (10%)
Each
Quantity:
30 x 60 cm
Request bulk or custom format
Price (USD)
287.65
Online Exclusive
319.00
Save 31.35 (10%)
Each
Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni:Mo; 68:17:12:3 wt%
Chemical Name or MaterialStainless Steel gauze, Type 316
MDL NumberMFCD00148499
Mesh Size200 Mesh
Name Note200 mesh woven from 0.05mm (0.002 in.) dia. wire
View more
Stainless Steel gauze is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only