Thermo Scientific Chemicals
Sapphire substrate, 50.8mm dia x 0.332mm thick, EPI polished two sides, C-axis, HEMCOR single crystal
Catalog number: 045020.KU
5 Ea., 50.8mm dia. x 0.332 mm thick, 0.332 mm, Each
Thermo Scientific Chemicals
Sapphire substrate, 50.8mm dia x 0.332mm thick, EPI polished two sides, C-axis, HEMCOR single crystal
Catalog number: 045020.KU
5 Ea., 50.8mm dia. x 0.332 mm thick, 0.332 mm, Each
Quantity
Specimen Size
Thickness
Catalog number: 045020.KU
also known as 045020-KU
Price (EUR)
Quantity
-
Comment
50.8mm dia x 0.332mm thick
Comment
C-axis