Polyethyleneimine, branched, M.W. 50,000 - 100,000, 30% w/w aq. soln.
Polyethyleneimine, branched, M.W. 50,000 - 100,000, 30% w/w aq. soln.
Polyethyleneimine, branched, M.W. 50,000 - 100,000, 30% w/w aq. soln.
Thermo Scientific Chemicals

Polyethyleneimine, branched, M.W. 50,000 - 100,000, 30% w/w aq. soln.

CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 045024.36
also known as 045024-36
Price (USD)
887.65
Online Exclusive
986.00
Save 98.35 (10%)
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
887.65
Online Exclusive
986.00
Save 98.35 (10%)
Each
Specifications
CAS9002-98-6
ChEBICHEBI:30969
Chemical Name or MaterialPolyethyleneimine
ColorColorless to Yellow
Concentration or Composition (by Analyte or Components)30% w/w aq. soln.
View more
Polyethyleneimine acts as a protein precipitant used to purify proteins. It is used as a chelating agent and as a scavenger for aldehydes and oxides. It is also used in detergents, paper industry, dyes, printing inks and in water treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only