Lead wire, 0.25mm (0.01 in.) dia., 99.95% (metals basis), Thermo Scientific Chemicals
Lead wire, 0.25mm (0.01 in.) dia., 99.95% (metals basis), Thermo Scientific Chemicals
Lead wire, 0.25mm (0.01 in.) dia., 99.95% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead wire, 0.25mm (0.01 in.) dia., 99.95% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
045025.G5
also known as 045025-G5
5 m
Catalog number 045025.G5
also known as 045025-G5
Price (USD)
416.65
Special offer
Online exclusive
Ends: 30-Jun-2026
490.00
Save 73.35 (15%)
Each
Quantity:
5 m
Request bulk or custom format
Price (USD)
416.65
Special offer
Online exclusive
Ends: 30-Jun-2026
490.00
Save 73.35 (15%)
Each
Chemical Identifiers
CAS7439-92-1
IUPAC Namelead
Molecular FormulaPb
InChI KeyWABPQHHGFIMREM-UHFFFAOYSA-N
SMILES[Pb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
FormWire, 0.25mm (0.01in) diameter.
Assay from Supplier's CofA≥99.95% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only