Sulfuric acid, 0.2N Standardized Solution
Sulfuric acid, 0.2N Standardized Solution
Sulfuric acid, 0.2N Standardized Solution
Sulfuric acid, 0.2N Standardized Solution
Thermo Scientific Chemicals

Sulfuric acid, 0.2N Standardized Solution

CAS: 7664-93-9 | H2O4S | 98.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 L
20 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045062.K7
also known as 045062-K7
Price (USD)
82.65
Online exclusive
92.00
Save 9.35
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
82.65
Online exclusive
92.00
Save 9.35
Each
Chemical Identifiers
CAS7664-93-9
IUPAC Namesulfuric acid
Molecular FormulaH2O4S
InChI KeyQAOWNCQODCNURD-UHFFFAOYSA-N
SMILESOS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA0.1996 N - 0.2004 N
FormLiquid
Sulfuric acid, 0.2N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only