Monel∣r 400 wire, 0.25mm (0.01in) dia
Thermo Scientific Chemicals

Monel∣r 400 wire, 0.25mm (0.01in) dia

Have Questions?
Change viewbuttonViewtableView
Quantity:
113 g
227 g
Catalog number 045065.TC
also known as 045065-TC
Price (USD)
115.65
Special offer
Online exclusive
Ends: 30-Jun-2026
136.00
Save 20.35 (15%)
Each
Quantity:
227 g
Request bulk or custom format
Price (USD)
115.65
Special offer
Online exclusive
Ends: 30-Jun-2026
136.00
Save 20.35 (15%)
Each
Specifications
Chemical Name or MaterialMonel™ 400 wire
MDL NumberMFCD09910536
Name Note0.25mm (0.01 in.) dia.
Recommended StorageAmbient temperatures
TSCAYes
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only