Palladium wire, 0.203mm (0.008in) dia, hard, 99.95% (metals basis)
Palladium wire, 0.203mm (0.008in) dia, hard, 99.95% (metals basis)
Palladium wire, 0.203mm (0.008in) dia, hard, 99.95% (metals basis)
Thermo Scientific Chemicals

Palladium wire, 0.203mm (0.008in) dia, hard, 99.95% (metals basis)

CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 m
10 m
Catalog number 045072.G6
also known as 045072-G6
Price (USD)
442.65
Special offer
Online exclusive
Ends: 30-Jun-2026
521.00
Save 78.35 (15%)
Each
Quantity:
10 m
Request bulk or custom format
Price (USD)
442.65
Special offer
Online exclusive
Ends: 30-Jun-2026
521.00
Save 78.35 (15%)
Each
Specifications
Assay Percent Notes(metals basis)
CAS5-3-7440
ChEBICHEBI:33363
Chemical Name or MaterialPalladium wire
EINECS Number231-115-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only