Tetraamminepalladium(II) hydrogen carbonate
Tetraamminepalladium(II) hydrogen carbonate
Tetraamminepalladium(II) hydrogen carbonate
Thermo Scientific Chemicals

Tetraamminepalladium(II) hydrogen carbonate

CAS: 134620-00-1 | C2H14N4O6Pd | 296.576 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 045082.03
also known as 045082-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS134620-00-1
IUPAC Namepalladium(2+) tetraamine dihydrogen carbonate
Molecular FormulaC2H14N4O6Pd
InChI KeyQYYPTZGPXKUKRM-UHFFFAOYSA-L
SMILESN.N.N.N.[Pd++].OC([O-])=O.OC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Purity≥97.0% (≥34.8% Pd)
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only