Polydimethylsiloxane (silicone) emulsion
Thermo Scientific Chemicals

Polydimethylsiloxane (silicone) emulsion

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 045088.36
also known as 045088-36
Price (USD)
72.10
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
72.10
Each
Specifications
Chemical Name or MaterialPolydimethylsiloxane (silicone) emulsion
MDL NumberMFCD00132673
Physical FormLiquid
Recommended StorageAmbient temperatures
Solubility InformationMiscible with carbon tetrachloride,benzene,chloroform,diethyl ether,toluene and other organic solvents. Immiscible with water and ethanol.
View more
Polydimethylsiloxane (silicone) emulsion is the most widely used silicon-based organic polymer and is particularly known for its unusual rheological (or flow) properties. Further, it is used in contact lenses, medical devices to elastomers and in shampoos (as dimethicone makes hair shiny and slippery). It finds application as an antifoaming agent in food, caulking, lubricants, kinetic sand and heat-resistant tiles. In addition to this, it serves as a critical ingredient in water-repelling coatings such as Rain-X.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only