Manganese powder, APS <10 micron, 99.6% (metals basis)
Manganese powder, APS <10 micron, 99.6% (metals basis)
Manganese powder, APS <10 micron, 99.6% (metals basis)
Thermo Scientific Chemicals

Manganese powder, APS <10 micron, 99.6% (metals basis)

CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 045098.18
also known as 045098-18
Price (USD)
39.80
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
39.80
Each
Chemical Identifiers
CAS7439-96-5
IUPAC Namemanganese
Molecular FormulaMn
InChI KeyPWHULOQIROXLJO-UHFFFAOYSA-N
SMILES[Mn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.4% max.
Appearance (Color)Gray
Particle SizeAPS < 10 microns
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only